siRNA Details
| Gene Symbol: Vmn2r124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271883.1 | NP_001258812.1 | 3 | 829 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r124| Gene Symbol: Vmn2r124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271883.1 | NP_001258812.1 | 3 | 829 | ||