siRNA Details
| Gene Symbol: Dusp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022019.5 | NP_071302.2 | 4 | 1352 | ||
| GenBank mRNA | AB037908.1 | - | 4 | 1358 | ||
| AK011995.1 | - | 4 | 1336 | |||
| AK012516.1 | - | 2 | 288 | |||
| AK035293.1 | - | 4 | 1350 | |||
| AK050528.1 | - | 4 | 1350 | |||
| AK088024.1 | - | 4 | 1351 | |||
| AK088186.1 | - | 4 | 1353 | |||
| AK088357.1 | - | 4 | 1348 | |||
| AK142568.1 | - | 4 | 1354 | |||
| BC025066.1 | - | 4 | 1370 | 3495358 | ||

Dusp10
Genomic Map