siRNA Details
| Gene Symbol: Gm7271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC147734.1 | - | 3 | 691 | 9056683 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm7271| Gene Symbol: Gm7271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC147734.1 | - | 3 | 691 | 9056683 | |