siRNA Details
| Gene Symbol: Gmfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022023.2 | NP_071306.2 | 6, 7 | 493 | ||
| GenBank mRNA | AB001732.1 | - | 6, 7 | 425 | ||
| AB050013.1 | - | 6, 7 | 435 | |||
| AF297220.1 | - | 6, 7 | 471 | |||
| AK013947.1 | - | 6, 7 | 450 | |||
| AK013970.1 | - | 6, 7 | 446 | |||
| AK142014.1 | - | 6, 7 | 465 | |||
| BC040233.1 | - | 6, 7 | 427 | 5364203 | ||
| BC049134.1 | - | 6, 7 | 434 | 6831501 | ||

Gmfb
Genomic Map