siRNA Details
| Gene Symbol: Tgm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160424.1 | NP_001153896.1 | 11 | 1902 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgm7| Gene Symbol: Tgm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160424.1 | NP_001153896.1 | 11 | 1902 | ||