siRNA Details
| Gene Symbol: Krt28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027574.1 | NP_081850.1 | 5 | 936 | ||
| GenBank mRNA | AB288231.1 | - | 5 | 936 | ||
| AK014642.1 | - | 5 | 937 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt28| Gene Symbol: Krt28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027574.1 | NP_081850.1 | 5 | 936 | ||
| GenBank mRNA | AB288231.1 | - | 5 | 936 | ||
| AK014642.1 | - | 5 | 937 | |||