siRNA Details
| Gene Symbol: Pinlyp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037143.2 | NP_001032220.1 | 3 | 257 | ||
| GenBank mRNA | AK009589.1 | - | 3 | 262 | ||
| AK009604.1 | - | 3 | 259 | |||
| AK009778.1 | - | 3 | 257 | |||
| AK009924.1 | - | 3 | 259 | |||
| BC139039.1 | - | 3 | 323 | 8862061 | ||
| BC145686.1 | - | 3 | 323 | 40130989 | ||

Pinlyp
Genomic Map