siRNA Details
| Gene Symbol: Fndc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022424.5 | NP_071869.2 | 3 | 576 | ||
| GenBank mRNA | AK013203.1 | - | 3 | 570 | ||
| AK018154.1 | - | 3 | 572 | |||
| AK163183.1 | - | 3 | 577 | |||
| BC139278.1 | - | 3 | 328 | 8862300 | ||
| BC139292.1 | - | 3 | 328 | 8862314 | ||

Fndc4
Genomic Map