siRNA Details
| Gene Symbol: Fndc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022424.5 | NP_071869.2 | 4, 5 | 808 | ||
| GenBank mRNA | AK013203.1 | - | 4, 5 | 802 | ||
| AK018154.1 | - | 4, 5 | 804 | |||
| AK163183.1 | - | 4, 5 | 809 | |||
| BC139278.1 | - | 4, 5 | 560 | 8862300 | ||
| BC139292.1 | - | 4, 5 | 560 | 8862314 | ||

Fndc4
Genomic Map