siRNA Details
| Gene Symbol: Ms4a4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029499.3 | NP_083775.1 | 2 | 228 | ||
| GenBank mRNA | AB026045.1 | - | 2 | 215 | ||
| AF237914.2 | - | 2 | 202 | |||
| AK017928.1 | - | 2 | 208 | |||
| AK151045.1 | - | 2 | 221 | |||
| AK153495.1 | - | 1 | 72 | |||
| AK157137.1 | - | 230 | ||||
| AK172650.1 | - | 2 | 200 | |||
| BC027496.1 | - | 2 | 184 | 1383416 | ||
| CT010170.1 | - | 1 | 96 | |||

Ms4a4c
Genomic Map