siRNA Details
| Gene Symbol: Sirt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122765.1 | NP_001116237.1 | 12 | 904 | ||
| NM_001122766.1 | NP_001116238.1 | 10 | 741 | |||
| NM_022432.4 | NP_071877.3 | 13 | 951 | |||
| GenBank mRNA | AB221570.1 | - | 13 | 856 | ||
| AF299337.1 | - | 13 | 936 | |||
| AK014042.1 | - | 12 | 875 | |||
| AK144431.1 | - | 12 | 848 | |||
| AK146412.1 | - | 12 | 881 | |||
| AK183534.1 | - | 5 | 325 | |||
| BC021439.1 | - | 13 | 955 | 5039278 | ||

Sirt2
Genomic Map