siRNA Details
| Gene Symbol: Sirt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127351.1 | NP_001120823.1 | 6, 7 | 999 | ||
| NM_001177804.1 | NP_001171275.1 | 6, 7 | 991 | |||
| NM_022433.2 | NP_071878.2 | 6, 7 | 1044 | |||
| GenBank mRNA | AF299338.1 | - | 6, 7 | 1044 | ||
| AF299339.1 | - | 6, 7 | 999 | |||
| AK075861.1 | - | 6, 7 | 973 | |||
| BC025878.1 | - | 6, 7 | 943 | 5253854 | ||
| BF681854.1 | - | 1, 2 | 225 | |||
| CT010402.1 | - | 5, 6 | 750 | |||
| EU886466.1 | - | 6, 7 | 989 | |||
| FJ621493.1 | - | 6, 7 | 987 | |||

Sirt3
Genomic Map