siRNA Details
| Gene Symbol: Polr1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285800.1 | NP_001272729.1 | 13 | 1264 | ||
| NM_022811.3 | NP_073722.1 | 11 | 1120 | |||
| GenBank mRNA | AK050780.1 | - | 1068 | |||
| AK083948.1 | - | 11 | 1090 | |||
| AK132556.1 | - | 11 | 1091 | |||
| D14336.1 | - | 11 | 1083 | |||

Polr1e
Genomic Map