siRNA Details
| Gene Symbol: Ireb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022655.3 | NP_073146.2 | 16 | 2100 | ||
| GenBank mRNA | BC044665.1 | - | 16 | 2067 | 2647641 | |
| CV558274.1 | - | 4 | 382 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ireb2| Gene Symbol: Ireb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022655.3 | NP_073146.2 | 16 | 2100 | ||
| GenBank mRNA | BC044665.1 | - | 16 | 2067 | 2647641 | |
| CV558274.1 | - | 4 | 382 | |||