siRNA Details
| Gene Symbol: Mrps25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025578.4 | NP_079854.2 | 4 | 549 | ||
| GenBank mRNA | AK004037.1 | - | 4 | 528 | ||
| AK013196.1 | - | 4 | 509 | |||
| AK131114.1 | - | 4 | 478 | |||
| BC022953.1 | - | 4 | 536 | 5126320 | ||
| CB595428.1 | - | 4 | 562 | |||

Mrps25
Genomic Map