siRNA Details
| Gene Symbol: Krtdap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033131.3 | NP_001028303.1 | 1 | 58 | ||
| GenBank mRNA | AK131603.1 | - | 1 | 59 | ||
| AK131606.1 | - | 1 | 59 | |||
| BC132510.1 | - | 1 | 58 | 40130787 | ||
| BC132512.1 | - | 1 | 58 | 40130789 | ||
| BC145437.1 | - | 1 | 58 | 9053995 | ||
| BC145438.1 | - | 1 | 58 | 9053996 | ||

Krtdap
Genomic Map