siRNA Details
Gene Symbol: Htra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019752.3 | NP_062726.3 | 5 | 1101 | ||
GenBank mRNA | AF164513.1 | - | 5 | 980 | ||
AF175324.1 | - | 5 | 997 | |||
AK011536.1 | - | 1907 | ||||
AK146338.1 | - | 5 | 992 | |||
AK159188.1 | - | 5 | 921 | |||
AK178298.1 | - | 3 | 84 | |||
AK182973.1 | - | 3 | 84 | |||
AK215600.1 | - | 3 | 84 | |||
BC033276.1 | - | 5 | 961 | |||
BC049880.1 | - | 5 | 1101 | 5321861 |
