siRNA Details
| Gene Symbol: Suv39h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022724.4 | NP_073561.2 | 3 | 741 | ||
| GenBank mRNA | AF149205.1 | - | 3 | 719 | ||
| AK083457.1 | - | 3 | 719 | |||
| AK165278.1 | - | 2 | 394 | |||
| BY752501.1 | - | 2 | 579 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Suv39h2| Gene Symbol: Suv39h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022724.4 | NP_073561.2 | 3 | 741 | ||
| GenBank mRNA | AF149205.1 | - | 3 | 719 | ||
| AK083457.1 | - | 3 | 719 | |||
| AK165278.1 | - | 2 | 394 | |||
| BY752501.1 | - | 2 | 579 | |||