siRNA Details
Gene Symbol: Vps35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022997.4 | NP_075373.1 | 15 | 1920 | ||
GenBank mRNA | AF226323.1 | - | 15 | 1939 | ||
AK135680.1 | - | 15 | 2232 | |||
AK142382.1 | - | 15 | 2207 | |||
AK145421.1 | - | 14 | 1889 | |||
AK145853.1 | - | 15 | 1968 | |||
AK150682.1 | - | 15 | 1934 | |||
AK162726.1 | - | 15 | 1899 | |||
AK166821.1 | - | 15 | 1913 | |||
AK167421.1 | - | 15 | 1937 | |||
AK167594.1 | - | 15 | 1933 | |||
AK168880.1 | - | 15 | 1913 | |||
AK170473.1 | - | 15 | 1954 | |||
AK187614.1 | - | 2 | 93 | |||
BC005469.1 | - | 15 | 1945 | 3492842 | ||
BC006637.1 | - | 15 | 1925 | 3969440 | ||
U47024.1 | - | 14 | 1894 |
