siRNA Details
| Gene Symbol: Bean1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141922.1 | NP_001135394.1 | 6 | 1199 | ||
| NM_001141924.1 | NP_001135396.1 | 4 | 982 | |||
| NM_001141925.1 | NP_001135397.1 | 4 | 984 | |||
| GenBank mRNA | AF240460.1 | - | 2 | 379 | ||
| AK134633.1 | - | 4 | 984 | |||
| BC030426.1 | - | 4 | 1062 | |||
| BC138184.1 | - | 6 | 1128 | 8861203 | ||

Bean1
Genomic Map