siRNA Details
| Gene Symbol: Tctn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039153.3 | NP_001034242.2 | 1 | 199 | ||
| GenBank mRNA | AK036568.1 | - | 1 | 181 | ||
| AK085676.1 | - | 1 | 159 | |||
| AK138661.1 | - | 1 | 198 | |||
| AK144772.1 | - | 1 | 175 | |||
| BC141084.1 | - | 1 | 154 | |||
| DQ278867.1 | - | 1 | 200 | |||

Tctn1
Genomic Map