siRNA Details
| Gene Symbol: Sdhc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025321.3 | NP_079597.2 | 5 | 376 | ||
| GenBank mRNA | AK002459.1 | - | 5 | 378 | ||
| AK012818.1 | - | 5 | 377 | |||
| AK032458.1 | - | 378 | ||||
| AK135503.1 | - | 5 | 378 | |||
| BC005779.1 | - | 5 | 363 | 2810852 | ||
| BC083091.1 | - | 5 | 369 | 30445339 | ||
| BY711932.1 | - | 5 | 377 | |||

Sdhc
Genomic Map