siRNA Details
| Gene Symbol: Krtcap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025327.2 | NP_079603.1 | 5 | 405 | ||
| GenBank mRNA | AK002479.1 | - | 5 | 406 | ||
| AK003310.1 | - | 5 | 373 | |||
| BC031492.1 | - | 5 | 358 | 3485092 | ||
| BC043030.1 | - | 5 | 877 | 5693076 | ||

Krtcap2
Genomic Map