siRNA Details
| Gene Symbol: Hsd17b14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025330.3 | NP_079606.3 | 9 | 859 | ||
| GenBank mRNA | AK002831.1 | - | 9 | 812 | ||
| AK010556.1 | - | 5 | 524 | |||
| BC064042.1 | - | 5 | 642 | 1332242 | ||
| BC099530.1 | - | 5 | 528 | |||

Hsd17b14
Genomic Map