siRNA Details
| Gene Symbol: Gtpbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159329.1 | NP_001152801.1 | 6 | 629 | ||
| NM_025332.3 | NP_079608.1 | 6 | 830 | |||
| GenBank mRNA | AK002771.1 | - | 6 | 815 | ||
| AK010979.1 | - | 6 | 827 | |||
| AK084919.1 | - | 1 | 830 | |||
| AK147840.1 | - | 6 | 814 | |||
| BC107216.1 | - | 6 | 827 | 40051675 | ||
| BC107217.1 | - | 6 | 626 | 40051680 | ||

Gtpbp8
Genomic Map