siRNA Details
| Gene Symbol: Rmnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025343.5 | NP_079619.4 | 9 | 1172 | ||
| GenBank mRNA | AK007323.1 | - | 9 | 1120 | ||
| AK075603.1 | - | 9 | 1155 | |||
| AK163523.1 | - | 408 | ||||
| AK205767.1 | - | 2 | 129 | |||
| AK215071.1 | - | 3 | 236 | |||
| AK215625.1 | - | 2 | 108 | |||
| BC019572.1 | - | 6 | 417 | |||
| BC027299.1 | - | 9 | 1120 | 3987394 | ||
| BC036150.1 | - | 8 | 1092 | 4977394 | ||

Rmnd1
Genomic Map