siRNA Details
| Gene Symbol: Rmnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025343.5 | NP_079619.4 | 5 | 873 | ||
| GenBank mRNA | AK007323.1 | - | 5 | 821 | ||
| AK075603.1 | - | 5 | 856 | |||
| AK163523.1 | - | 109 | ||||
| BC019572.1 | - | 2 | 118 | |||
| BC027299.1 | - | 5 | 821 | 3987394 | ||
| BC036150.1 | - | 4 | 793 | 4977394 | ||

Rmnd1
Genomic Map