siRNA Details
Gene Symbol: Eif3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025344.2 | NP_079620.2 | 2, 3 | 469 | ||
GenBank mRNA | AK002778.1 | - | 2, 3 | 468 | ||
AK003372.1 | - | 2, 3 | 467 | |||
AK010333.1 | - | 2, 3 | 463 | |||
AK088537.1 | - | 2, 3 | 469 | |||
AK136797.1 | - | 468 | ||||
AK152037.1 | - | 2, 3 | 470 | |||
AK190701.1 | - | 2, 3 | 114 | |||
AK197956.1 | - | 2, 3 | 116 | |||
AK217928.1 | - | 1, 2 | 87 | |||
BC070473.1 | - | 2, 3 | 451 | 30634857 | ||
BC083190.1 | - | 2, 3 | 454 | 30358339 |
