siRNA Details
Gene Symbol: Eif3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025344.2 | NP_079620.2 | 6 | 814 | ||
GenBank mRNA | AK002778.1 | - | 6 | 813 | ||
AK003372.1 | - | 6 | 812 | |||
AK088537.1 | - | 6 | 814 | |||
AK136797.1 | - | 813 | ||||
AK152037.1 | - | 6 | 815 | |||
AY061808.1 | - | 3 | 158 | |||
BC070473.1 | - | 6 | 796 | 30634857 | ||
BC083190.1 | - | 6 | 799 | 30358339 |
