siRNA Details
| Gene Symbol: Ypel3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025347.2 | NP_079623.1 | 4 | 419 | ||
| NM_026875.2 | NP_081151.2 | 4 | 519 | |||
| GenBank mRNA | AB098742.1 | - | 3 | 224 | ||
| AF523353.1 | - | 4 | 409 | |||
| AK002925.1 | - | 4 | 404 | |||
| AK003371.1 | - | 4 | 420 | |||
| AK004431.1 | - | 4 | 492 | |||
| BC009171.1 | - | 4 | 393 | |||

Ypel3
Genomic Map