siRNA Details
Gene Symbol: Ndufa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025348.2 | NP_079624.1 | 3, 4 | 188 | ||
GenBank mRNA | AK003133.1 | - | 3, 4 | 179 | ||
AK019099.1 | - | 3, 4 | 190 | |||
BC061002.1 | - | 181 | ||||
BC120712.1 | - | 3, 4 | 182 | 40129635 | ||
BC120714.1 | - | 3, 4 | 182 | 40129637 |
