siRNA Details
| Gene Symbol: Ndufa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025348.2 | NP_079624.1 | 4 | 258 | ||
| GenBank mRNA | AK003133.1 | - | 4 | 249 | ||
| AK019099.1 | - | 4 | 260 | |||
| BC061002.1 | - | 251 | ||||
| BC120712.1 | - | 4 | 252 | 40129635 | ||
| BC120714.1 | - | 4 | 252 | 40129637 | ||

Ndufa3
Genomic Map