siRNA Details
Gene Symbol: Ghitm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199122.1 | NP_001186051.1 | 5 | 622 | ||
NM_078478.5 | NP_510963.1 | 5 | 575 | |||
GenBank mRNA | AF412297.1 | - | 5 | 519 | ||
AK005636.1 | - | 5 | 519 | |||
AK093887.1 | - | 5 | 551 | |||
AK134592.1 | - | 5 | 505 | |||
AK146433.1 | - | 5 | 482 | |||
AK146853.1 | - | 5 | 538 | |||
AK151488.1 | - | 5 | 552 | |||
AK158904.1 | - | 5 | 565 | |||
AK159695.1 | - | 5 | 549 | |||
AK159724.1 | - | 5 | 549 | |||
AK164122.1 | - | 5 | 520 | |||
AK167970.1 | - | 5 | 546 | |||
AK168022.1 | - | 5 | 552 | |||
AK168678.1 | - | 5 | 623 | |||
AK168800.1 | - | 5 | 531 | |||
BC008622.1 | - | 5 | 553 | 3588758 | ||
BC010224.1 | - | 5 | 555 | 3500685 |
