siRNA Details
| Gene Symbol: Wfdc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183249.2 | NP_899072.1 | 1 | 132 | ||
| GenBank mRNA | AA254806.1 | - | 1 | 62 | ||
| AK003164.1 | - | 1 | 74 | |||
| AK003352.1 | - | 1 | 76 | |||
| BC096415.1 | - | 1 | 67 | |||
| BY082582.1 | - | 1 | 132 | |||
| BY094209.1 | - | 1 | 74 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK155780.1 | - | 74 | Chr. - - [-] on Build | ||

Wfdc21
Genomic Map