siRNA Details
| Gene Symbol: Ndufa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025358.3 | NP_079634.2 | 4 | 406 | ||
| GenBank mRNA | AK003137.1 | - | 4 | 388 | ||
| AK003214.1 | - | 4 | 379 | |||
| AK012366.1 | - | 4 | 386 | |||
| BC005760.1 | - | 3 | 287 | |||
| BC058378.1 | - | 4 | 374 | 6820787 | ||

Ndufa9
Genomic Map