siRNA Details
| Gene Symbol: Tmem8c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159602.1 | NP_001153074.1 | 2 | 144 | ||
| NM_025376.3 | NP_079652.1 | 2 | 296 | |||
| GenBank mRNA | AK003298.1 | - | 2 | 298 | ||
| AK004477.1 | - | 2 | 265 | |||
| BB839715.1 | - | 2 | 298 | |||
| BC062145.1 | - | 2 | 222 | 30286808 | ||
| CX564956.1 | - | 2 | 145 | |||

Tmem8c
Genomic Map