siRNA Details
Gene Symbol: Necap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025383.3 | NP_079659.1 | 1, 2 | 129 | ||
GenBank mRNA | AK003463.1 | - | 1, 2 | 96 | ||
AK145874.1 | - | 1, 2 | 117 | |||
AK155871.1 | - | 1, 2 | 117 | |||
AK156566.1 | - | 1, 2 | 106 | |||
AK170987.1 | - | 1, 2 | 114 | |||
AK172538.1 | - | 1, 2 | 99 | |||
AK197123.1 | - | 21 | ||||
AK213029.1 | - | 1, 2 | 21 | |||
AK218572.1 | - | 1, 2 | 38 | |||
BB851186.1 | - | 1, 2 | 128 | |||
BC037069.1 | - | 1, 2 | 128 | 5002765 |
