siRNA Details
| Gene Symbol: Bola2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175103.3 | NP_780312.1 | 1, 2 | 109 | ||
| GenBank mRNA | AK002662.1 | - | 1, 2 | 112 | ||
| AK003922.1 | - | 1, 2 | 112 | |||
| AK028165.1 | - | 1, 2 | 123 | |||
| BC045523.1 | - | 1, 2 | 127 | 6397886 | ||
| BC061197.1 | - | 1, 2 | 137 | 6707992 | ||

Bola2
Genomic Map