siRNA Details
| Gene Symbol: Ubl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025401.3 | NP_079677.1 | 2 | 1622 | ||
| GenBank mRNA | AK002730.1 | - | 2 | 131 | ||
| AK003992.1 | - | 2 | 138 | |||
| AK007726.1 | - | 2 | 103 | |||
| AK009854.1 | - | 2 | 138 | |||
| AK012803.1 | - | 2 | 120 | |||
| AK051149.1 | - | 2 | 1623 | |||
| AK154036.1 | - | 2 | 2068 | |||
| BC028498.1 | - | 2 | 92 | 1400865 | ||

Ubl5
Genomic Map