siRNA Details
| Gene Symbol: 1110059G10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025419.4 | NP_079695.2 | 3 | 363 | ||
| GenBank mRNA | AK004312.1 | - | 3 | 359 | ||
| AK012115.1 | - | 3 | 359 | |||
| AK083316.1 | - | 3 | 336 | |||
| AK160617.1 | - | 3 | 364 | |||
| AK220157.1 | - | 2 | 171 | |||
| BC034133.1 | - | 3 | 321 | 4921846 | ||

1110059G10Rik
Genomic Map