siRNA Details
| Gene Symbol: Cd302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025422.4 | NP_079698.2 | 4, 5 | 822 | ||
| GenBank mRNA | AK004267.1 | - | 4, 5 | 758 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd302| Gene Symbol: Cd302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025422.4 | NP_079698.2 | 4, 5 | 822 | ||
| GenBank mRNA | AK004267.1 | - | 4, 5 | 758 | ||