siRNA Details
| Gene Symbol: Inip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013577.1 | NP_001013595.1 | 3 | 278 | ||
| GenBank mRNA | AK004256.1 | - | 2 | 151 | ||
| AK005245.1 | - | 3 | 260 | |||
| AK012023.1 | - | 3 | 198 | |||
| AK159116.1 | - | 3 | 266 | |||
| AK169537.1 | - | 2 | 180 | |||
| BB641174.1 | - | 3 | 278 | |||
| BC116204.1 | - | 3 | 173 | 40090152 | ||
| BC116205.1 | - | 3 | 173 | 40090155 | ||

Inip
Genomic Map