siRNA Details
| Gene Symbol: Rgcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025427.2 | NP_079703.2 | 3, 4 | 487 | ||
| GenBank mRNA | AF276981.1 | - | 3, 4 | 499 | ||
| AK004451.1 | - | 3, 4 | 488 | |||
| AK004705.1 | - | 3, 4 | 483 | |||
| BC049580.1 | - | 3, 4 | 483 | 6741281 | ||
| BC050935.1 | - | 3, 4 | 476 | 6818687 | ||
| CK792184.1 | - | 2, 3 | 451 | |||
| CT010403.1 | - | 3, 4 | 349 | |||

Rgcc
Genomic Map