siRNA Details
Gene Symbol: Ing5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025454.2 | NP_079730.1 | 6 | 595 | ||
GenBank mRNA | AK006421.1 | - | 6 | 694 | ||
AK007536.1 | - | 6 | 586 | |||
AK028010.1 | - | 6 | 573 | |||
AK028397.1 | - | 6 | 596 | |||
AK145695.1 | - | 6 | 568 | |||
BC064674.1 | - | 6 | 594 | 6400183 |
