siRNA Details
| Gene Symbol: Tmem126a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025460.2 | NP_079736.1 | 3 | 250 | ||
| GenBank mRNA | AK005343.1 | - | 3 | 229 | ||
| AK007559.1 | - | 3 | 251 | |||
| AK050267.1 | - | 3 | 243 | |||
| BC023171.1 | - | 3 | 214 | 4946998 | ||
| EF437357.1 | - | 2 | 66 | |||

Tmem126a
Genomic Map