siRNA Details
| Gene Symbol: Aamdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177945.1 | NP_001171416.1 | 4 | 451 | ||
| NM_183251.3 | NP_899074.2 | 3 | 347 | |||
| GenBank mRNA | AA065573.1 | - | 3 | 284 | ||
| AW124043.1 | - | 2 | 218 | |||
| BC026557.1 | - | 4 | 369 | 5027461 | ||

Aamdc
Genomic Map