siRNA Details
| Gene Symbol: Smim20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145433.1 | NP_001138905.1 | 1 | 176 | ||
| GenBank mRNA | AK003139.1 | - | 1 | 111 | ||
| AK007474.1 | - | 1 | 99 | |||
| AK156339.1 | - | 1 | 92 | |||
| BC030079.1 | - | 1 | 112 | |||
| BY077818.1 | - | 1 | 177 | |||

Smim20
Genomic Map