siRNA Details
Gene Symbol: Dpy30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146222.1 | NP_001139694.1 | 4 | 394 | ||
NM_001146223.1 | NP_001139695.1 | 4 | 308 | |||
NM_001146224.1 | NP_001139696.1 | 3 | 314 | |||
NM_024428.4 | NP_077746.3 | 4 | 403 | |||
GenBank mRNA | AK003688.1 | - | 3 | 239 | ||
AK011643.1 | - | 3 | 213 | |||
AK013073.1 | - | 4 | 297 | |||
AK077902.1 | - | 4 | 296 | |||
AK214978.1 | - | 3 | 186 | |||
AV460563.1 | - | 4 | 315 | |||
AY297538.1 | - | 4 | 407 | |||
BC002240.1 | - | 4 | 277 | 3491912 | ||
BU961422.1 | - | 4 | 409 | |||
BX518196.1 | - | 4 | 409 |
