siRNA Details
| Gene Symbol: Tmem208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025486.2 | NP_079762.1 | 5 | 472 | ||
| GenBank mRNA | AK002499.1 | - | 5 | 460 | ||
| AK005662.1 | - | 5 | 456 | |||
| BC005665.1 | - | 5 | 427 | |||
| BC024608.1 | - | 5 | 423 | 4242513 | ||
| BE983091.1 | - | 2 | 316 | |||

Tmem208
Genomic Map