siRNA Details
| Gene Symbol: Riiad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025506.2 | NP_079782.1 | 2 | 139 | ||
| GenBank mRNA | AK009192.1 | - | 2 | 141 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Riiad1| Gene Symbol: Riiad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025506.2 | NP_079782.1 | 2 | 139 | ||
| GenBank mRNA | AK009192.1 | - | 2 | 141 | ||